Learn More
Cayman Chemical Precellys 2 mL Soft Tissue H
SDP
Catalog No. 502886944
Encompass_Preferred
Kit Includes 50 preps prefilled with 1.4 mm ceramic (zirconium oxide) beads in 2 mL standard tubes. CK14 is designed for soft tissue homogenization like brain, liver, kidney, skin, plant leaves, mammalian cells, and more. Sample size range from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000912-LYSK0-A.0; Previously KT03961-1-003.2]
Catalog No. 50-288-6944 Supplier Cayman Chemical Supplier No. 100111521 BOX
Kit Includes 50 preps prefilled with 1.4 mm ceramic (zirconium oxide) beads in 2 mL standard tubes. CK14 is designed for soft tissue homogenization like brain, liver, kidney, skin, plant leaves, mammalian cells, and more. Sample size range from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000912-LYSK0-A.0; Previously KT03961-1-003.2]