Learn More
Cayman Chemical Precellys 2 mL Soil Grinding
SDP
Supplier:  Cayman Chemical 100111951 BOX
Encompass_Preferred
Kit includes 50 preps of a mix of glass and ceramic (zirconium oxide) beads in 2 mL standard tubes. SK38 is designed for micro-organism lysing included in samples like soil, sediment, roots,feces, animal or plant tissues, and more. Sample size range from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000915-LYSK0-A.0; Previously KT03961-1-006.2]
Catalog No. 50-288-6612