Learn More
Cayman Chemical Precellys 2 mL Tissue Grindi
SDP
Catalog No. 502972848
Encompass_Preferred
Kit includes 50 preps of ceramic (zirconium oxide) mix beads of 2.8 mm and 5.0 mm in 2 mL reinforced tubes. Custom design reinforced tubes for bead beating application: O-ring and reinforced materials for dry grinding, valuable or very hard samples. CKMix50 Reinforced is designed for wet or dry tissue grinding like hair, whole piece of mouse lung, heart or dry hair, mouse bones and more. Sample sizes range from 20 mg to 200 mg tissue or 200 µL to 1. 6 mL culture. [Bertin Catalog P000922-LYSK0-A.0; Previously KT03961-1-013.2]
Catalog No. 50-297-2848 Supplier Cayman Chemical Supplier No. 152342 BOX
Kit includes 50 preps of ceramic (zirconium oxide) mix beads of 2.8 mm and 5.0 mm in 2 mL reinforced tubes. Custom design reinforced tubes for bead beating application: O-ring and reinforced materials for dry grinding, valuable or very hard samples. CKMix50 Reinforced is designed for wet or dry tissue grinding like hair, whole piece of mouse lung, heart or dry hair, mouse bones and more. Sample sizes range from 20 mg to 200 mg tissue or 200 µL to 1. 6 mL culture. [Bertin Catalog P000922-LYSK0-A.0; Previously KT03961-1-013.2]