Learn More
Cayman Chemical Precellys 2 mL Tissue Homoge
SDP
Catalog No. 502886861
Encompass_Preferred
Kit includes 50 preps of ceramic (zirconium oxide) mix beads of 1.4 mm and 2.8 mm in 2 ml standard tubes. CKMix is designed for tissue homogenization like brain, liver, kidney, EAr, lung, heart, spleen, tail, artery, arabidopsis, leaves, tumor, intestine, adipose tissue and more. Sample size range from 20 mg to 200 mg tissue or 200 µl to 1.6 ml culture. [Bertin Catalog P000918-LYSK0-A.00; Previously KT03961-1-009.2]
Catalog No. 50-288-6861 Supplier Cayman Chemical Supplier No. 1040910 BOX
Kit includes 50 preps of ceramic (zirconium oxide) mix beads of 1.4 mm and 2.8 mm in 2 ml standard tubes. CKMix is designed for tissue homogenization like brain, liver, kidney, EAr, lung, heart, spleen, tail, artery, arabidopsis, leaves, tumor, intestine, adipose tissue and more. Sample size range from 20 mg to 200 mg tissue or 200 µl to 1.6 ml culture. [Bertin Catalog P000918-LYSK0-A.00; Previously KT03961-1-009.2]