Learn More
Cayman Chemical Precellys 2 mL Tissue Homoge
Supplier: Cayman Chemical 104091 BOX
Kit includes 50 preps of ceramic (zirconium oxide) mix beads of 1.4 mm and 2.8 mm in 2 ml standard tubes. CKMix is designed for tissue homogenization like brain, liver, kidney, EAr, lung, heart, spleen, tail, artery, arabidopsis, leaves, tumor, intestine, adipose tissue and more. Sample size range from 20 mg to 200 mg tissue or 200 µl to 1.6 ml culture. [Bertin Catalog P000918-LYSK0-A.00; Previously KT03961-1-009.2]