Learn More
Bertin Corp Precellys 24 Touch Homogenizer
SDP
Catalog No. NC2063622
Encompass_Preferred
Precellys 24 Touch is the ideal lab instrument to homogenize any kind of biological sample, from the softest to the hardest, regardless of its origin (animal tissues, plant tissues, microorganisms¿). It can treat up to 24 samples at a time and has been validated on more than 1,500 different samples: from brain, muscle and heart samples, to very hard material such as bones, hair and seeds.
Catalog No. NC2063622 Supplier Bertin Corp Supplier No. P002391P24T0A.0
Add to Cart
Add to Cart
Request a Quote
Precellys 24 Touch is the ideal lab instrument to homogenize any kind of biological sample, from the softest to the hardest, regardless of its origin (animal tissues, plant tissues, microorganisms¿). It can treat up to 24 samples at a time and has been validated on more than 1,500 different samples: from brain, muscle and heart samples, to very hard material such as bones, hair and seeds.