Learn More
Cayman Chemical Precellys 24 White Blocking
SDP
Catalog No. 502886636
Encompass_Preferred
This spare part is designed for the Precellys 24 Tissue Homogenizer. This part will block the tubes on the holder with the vacuum technology, in order to provide a total security for both end-user and samples. [Bertin Catalog S000680-PR240-A.0; Previously SP03119-810-NC002.0]
Catalog No. 50-288-6636 Supplier Cayman Chemical Supplier No. 104221 EA
This spare part is designed for the Precellys 24 Tissue Homogenizer. This part will block the tubes on the holder with the vacuum technology, in order to provide a total security for both end-user and samples. [Bertin Catalog S000680-PR240-A.0; Previously SP03119-810-NC002.0]