Learn More
Cayman Chemical Precellys 7 mL Bacteria Lysi
SDP
Catalog No. 502955774
Encompass_Preferred
Kit includes 50 preps of 0.1 mm ceramic (zirconium oxide) beads in 7 ml standard tubes with O-ring. CK01 7 ml is designed for micro-organism lysing like gram+, gram- bacteria, spores and more. Sample size range: from 200 mg to 2 g sample or 200 µl to 6 ml culture. [Bertin Catalog P000938-LYSK0-A.0; Previously KT03961-1-305.7]
Catalog No. 50-295-5774 Supplier Cayman Chemical Supplier No. 137641 BOX
Kit includes 50 preps of 0.1 mm ceramic (zirconium oxide) beads in 7 ml standard tubes with O-ring. CK01 7 ml is designed for micro-organism lysing like gram+, gram- bacteria, spores and more. Sample size range: from 200 mg to 2 g sample or 200 µl to 6 ml culture. [Bertin Catalog P000938-LYSK0-A.0; Previously KT03961-1-305.7]