Learn More
Cayman Chemical Precellys 7 mL Hard Tissue H
SDP
Supplier:  Cayman Chemical 104081 BOX
Encompass_Preferred
Kit includes 50 preps of 2.8 mm ceramic (zirconium oxide) beads in 7 ml standard tubes with O-ring. CK28 7 ml is designed for tissue homogenization like whole rat brain, whole liver, kidney, spleen, leaves and more. Sample size range from 200 mg to 2 g tissue or 200 µl to 6 ml culture. [Bertin Catalog P000935-LYSK0-A.0; Previously KT03961-1-302.7]
Catalog No. 50-288-6849