Learn More
Cayman Chemical Precellys 7 mL Soil Grinding
SDP
Supplier:  Cayman Chemical 137621 BOX
Encompass_Preferred
Kit includes 50 preps of a mix of ceramic (zirconium oxide) and glass beads in 7 mL standard tubes with O-ring. SK38 7 mL is designed for micro-organism lysing included in samples like soil, sediment, root, feces, animal or plant tissues and more. Sample size range from 200 mg to 2 g tissue or 200 µL to 6 mL culture. [Bertin Catalog P000936-LYSK0-A.0; Previously KT03961-1-303.7]
Catalog No. 50-295-5775