Learn More
Cayman Chemical Precellys 7 mL Tissue Grindi
SDP
Catalog No. 502971247
Encompass_Preferred
Kit includes 50 preps of a mix of ceramic (zirconium oxide) mix beads of 2.8 mm and 5.0 mm in 7 mL standard tubes with O-ring. The 7 mL CKMix50 is designed for grinding whole organs like rat brain, liver, kidney, skin, whole iris-ciliary body pork, and dry samples as pepper grains and more. Sample size range from 200 mg to 2 g tissue or 200 µL to 6 mL culture. [Bertin Catalog P000939-LYSK0-A.0; Previously KT03961-1-306.7]
Catalog No. 50-297-1247 Supplier Cayman Chemical Supplier No. 152375 BOX
Kit includes 50 preps of a mix of ceramic (zirconium oxide) mix beads of 2.8 mm and 5.0 mm in 7 mL standard tubes with O-ring. The 7 mL CKMix50 is designed for grinding whole organs like rat brain, liver, kidney, skin, whole iris-ciliary body pork, and dry samples as pepper grains and more. Sample size range from 200 mg to 2 g tissue or 200 µL to 6 mL culture. [Bertin Catalog P000939-LYSK0-A.0; Previously KT03961-1-306.7]