Learn More
Cayman Chemical Precellys 7 mL Tube Centrifu
SDP
Catalog No. 502954240
Encompass_Preferred
The 7 mL Tube Centrifuge Adapter Pack contains 2 cetrifuge adapters. These adapters are designed to be used in centrifuges that can handle 15 mL tubes. The adapter fits in the 15 mL tube cavity. Precellys 7 mL tubes can now be EAsily spun down. [Bertin Catalog S000749-PR480-A.0; Previously SP03961-810-NC002.0]
Catalog No. 50-295-4240 Supplier Cayman Chemical Supplier No. 139361 EA
The 7 mL Tube Centrifuge Adapter Pack contains 2 cetrifuge adapters. These adapters are designed to be used in centrifuges that can handle 15 mL tubes. The adapter fits in the 15 mL tube cavity. Precellys 7 mL tubes can now be EAsily spun down. [Bertin Catalog S000749-PR480-A.0; Previously SP03961-810-NC002.0]