Learn More
Bertin Corp Precellys Emulsion Kit
SDP
Catalog No. NC2065743
Encompass_Preferred
The Precellys Emulsion Kit provides a reliable and rapid method to generate injectable antigen or antibody emulsions in the context of immunological research.
Catalog No. NC2065743 Supplier Bertin Corp Supplier No. D34200.10 EA
The Precellys Emulsion Kit provides a reliable and rapid method to generate injectable antigen or antibody emulsions in the context of immunological research.