Learn More
Bertin Corp Centrifuge adapters for 7mL Precellys Lysing kits
SDP
Catalog No. 501542979
Encompass_Preferred
Centrifuge adapters specially designed to centrifuge the 7mL Precellys lysing tubes. Adpaters fit in the standard 50mL tube holes of centrifuges.
Catalog No. 50-154-2979 Supplier Bertin Corp Supplier No. S000749PR480A.0
Centrifuge adapters specially designed to centrifuge the 7mL Precellys lysing tubes. Adpaters fit in the standard 50mL tube holes of centrifuges.