Learn More
Cayman Chemical Precellys Glass Beads 0 1 m
SDP
Catalog No. 502886842
Encompass_Preferred
Includes 500 preps of 0.1 mm glass beads. These beads are designed for use with Precellys Tissue Homogenizers. Beads are designed for micro-organism lysing like human cells, gram+, bacteria, and more. [Bertin Catalog P000929-LYSK0-A.0; Previously KT03961-1-105.BK]
Catalog No. 50-288-6842 Supplier Cayman Chemical Supplier No. 104041 BOX
Includes 500 preps of 0.1 mm glass beads. These beads are designed for use with Precellys Tissue Homogenizers. Beads are designed for micro-organism lysing like human cells, gram+, bacteria, and more. [Bertin Catalog P000929-LYSK0-A.0; Previously KT03961-1-105.BK]