Learn More
Cayman Chemical Precellys Glass Beads 0 5 m
SDP
Catalog No. 502886875
Encompass_Preferred
Includes 500 preps of 0.5 mm glass beads. These beads are designed for use with Precellys Tissue Homogenizers. Beads are designed for tough micro-organism lysing like human cells, gram+, gram-, bacteria, and more. [Bertin Catalog P000928-LYSK0-A.0; Previously KT03961-1-104.BK]
Catalog No. 50-288-6875 Supplier Cayman Chemical Supplier No. 104031 BOX
Includes 500 preps of 0.5 mm glass beads. These beads are designed for use with Precellys Tissue Homogenizers. Beads are designed for tough micro-organism lysing like human cells, gram+, gram-, bacteria, and more. [Bertin Catalog P000928-LYSK0-A.0; Previously KT03961-1-104.BK]