Learn More
Bertin Corp 0.5mm Glass beads (400g)
SDP
Catalog No. 501542946
Encompass_Preferred
Beads in bulk containing 400g of 0.5mm diameter Glass beads indicated for micro-organism gram-, gram+ bacteria, fungal spores and walls. To be combined with Precellys empty tubes.
Catalog No. 50-154-2946 Supplier Bertin Corp Supplier No. P000928LYSK0A.0
Beads in bulk containing 400g of 0.5mm diameter Glass beads indicated for micro-organism gram-, gram+ bacteria, fungal spores and walls. To be combined with Precellys empty tubes.