Learn More
Bertin Corp 0.1mm Glass beads (400g)
SDP
Catalog No. 501542947
Encompass_Preferred
Beads in bulk containing 400g of 0.1mm diameter glass beads of 0.1mm.0.1mm glass beads are indicated for micro-organism lysing like gram-, gram+ bacteria, fungal spores and walls. To be combined with Precellys empty tubes.
Catalog No. 50-154-2947 Supplier Bertin Corp Supplier No. P000929LYSK0A.0
Beads in bulk containing 400g of 0.1mm diameter glass beads of 0.1mm.0.1mm glass beads are indicated for micro-organism lysing like gram-, gram+ bacteria, fungal spores and walls. To be combined with Precellys empty tubes.