Learn More
Bertin Corp 2.8mm Stainless steel beads (500g)
SDP
Catalog No. 501542943
Encompass_Preferred
Beads in bulk, containing 500g of 2.8mm diameter Stainless steel beads. To be combined with Precellys empty tubes.
Catalog No. 50-154-2943 Supplier Bertin Corp Supplier No. P000925LYSK0A.0
Beads in bulk, containing 500g of 2.8mm diameter Stainless steel beads. To be combined with Precellys empty tubes.