Learn More
Bertin Corp Bacteria Lysing CK01 - 15mL tubes 25 prep
SDP
Catalog No. 501542964
Encompass_Preferred
Lysing kit for Precellys - CK01 Bacteria kit - 15mL tubes 25 prep. The CK01 matrix is composed of 0.1mm ceramic (zirconium oxide) beads and is designed for micro-organisms lysing like gram- and gram+ bacteria but also spores, micro-algae etc. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 2g to 4g of tissue. The kits requires the use of the dedicated 15mL holder pack for Precellys Evolution (SP000810-PEVO0-A.0)
Catalog No. 50-154-2964 Supplier Bertin Corp Supplier No. P000948LYSK0A.0
Lysing kit for Precellys - CK01 Bacteria kit - 15mL tubes 25 prep. The CK01 matrix is composed of 0.1mm ceramic (zirconium oxide) beads and is designed for micro-organisms lysing like gram- and gram+ bacteria but also spores, micro-algae etc. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 2g to 4g of tissue. The kits requires the use of the dedicated 15mL holder pack for Precellys Evolution (SP000810-PEVO0-A.0)