Learn More
Bertin Corp Dry Hard Tissue Grinding CK68 - 15mL tubes 25 Prep
SDP
Catalog No. 501542966
Encompass_Preferred
Lysing kit for Precellys - CK68 Dry Hard Tissue kit - 15mL tubes 25 prep. The CK68 matrix is composed of 6.8mmceramic (zirconium oxide) bead recommended for hard tissues such as muscle, bone, teeth, hair but also for plant roots, rods or seeds like corn, rice, wheat etc. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 2g to 4g of tissue. The kits requires the use of the dedicated tube holder for Precellys Evolution (S000810-PEVO0-A.0).
Catalog No. 50-154-2966 Supplier Bertin Corp Supplier No. P000950LYSK0A.0
Lysing kit for Precellys - CK68 Dry Hard Tissue kit - 15mL tubes 25 prep. The CK68 matrix is composed of 6.8mmceramic (zirconium oxide) bead recommended for hard tissues such as muscle, bone, teeth, hair but also for plant roots, rods or seeds like corn, rice, wheat etc. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 2g to 4g of tissue. The kits requires the use of the dedicated tube holder for Precellys Evolution (S000810-PEVO0-A.0).