Learn More
Bertin Corp Hard Tissue Homogenizing CK28 - 2mL tubes 50 Prep
SDP
Catalog No. 501542931
Encompass_Preferred
Lysing kit for Precellys - CK28 Hard tissue - 2mL tubes 50 prep. The CK28 matrix is composed of 2.8mm Ceramic (zirconium oxide) beads recommended for hard tissues such as heart, muscle, skin, spinal cord, intestine, adipose tissues but also for plant roots, rods or seeds like corn, rice, wheat... It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24
Catalog No. 50-154-2931 Supplier Bertin Corp Supplier No. P000911LYSK0A.0
Lysing kit for Precellys - CK28 Hard tissue - 2mL tubes 50 prep. The CK28 matrix is composed of 2.8mm Ceramic (zirconium oxide) beads recommended for hard tissues such as heart, muscle, skin, spinal cord, intestine, adipose tissues but also for plant roots, rods or seeds like corn, rice, wheat... It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24