Learn More
Bertin Corp Tissue Grinding CK50-R - 2mL Reinforced tubes 50 Prep
SDP
Catalog No. 501542940
Encompass_Preferred
Reinforced Lysing kit for Precellys - CKMix50-R Hard tissue kit -2mL tubes 50 prep. The CKMix50 matrix is composed of a dual ceramic (zirconium oxide) bead type of 2.8mm and 5.0mm in reinforced tubes and is recommended for hard tissues such as bones, teeth, hair. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 200mg to 2g of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24.
Catalog No. 50-154-2940 Supplier Bertin Corp Supplier No. P000922LYSK0A.0
Reinforced Lysing kit for Precellys - CKMix50-R Hard tissue kit -2mL tubes 50 prep. The CKMix50 matrix is composed of a dual ceramic (zirconium oxide) bead type of 2.8mm and 5.0mm in reinforced tubes and is recommended for hard tissues such as bones, teeth, hair. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 200mg to 2g of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24.