Learn More
Bertin Corp Soft Tissue Homogenizing CK14 - 2mL tubes 50 Prep
SDP
Catalog No. 501542932
Encompass_Preferred
Lysing kit for Precellys - CK 14 Soft tissue - 2mL 50 prep. The CK14 is a ceramic (zirconium oxide) matrix composed of 1.4mm beads and is indicated in the homogenization of soft tissues such as liver, kidney, brain, but also on plants parts such as leaves. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24
Catalog No. 50-154-2932 Supplier Bertin Corp Supplier No. P000912LYSK0A.0
Lysing kit for Precellys - CK 14 Soft tissue - 2mL 50 prep. The CK14 is a ceramic (zirconium oxide) matrix composed of 1.4mm beads and is indicated in the homogenization of soft tissues such as liver, kidney, brain, but also on plants parts such as leaves. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24