Learn More
Bertin Corp Tough Micro-organism Lysing VK05 - 2mL tubes 50 Prep
SDP
Catalog No. 501542933
Encompass_Preferred
Lysing kit for Precellys - VK05 Tough Micro-organisms - 2mL 50 prep. The VK05 matrix is composed of 0.5mm glass beads and is designed for tough micro-organism lysing like yeast, fungi, cyanobacteria, phytoplankton, nematode, spores, micro-algae and more. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24
Catalog No. 50-154-2933 Supplier Bertin Corp Supplier No. P000913LYSK0A.0
Lysing kit for Precellys - VK05 Tough Micro-organisms - 2mL 50 prep. The VK05 matrix is composed of 0.5mm glass beads and is designed for tough micro-organism lysing like yeast, fungi, cyanobacteria, phytoplankton, nematode, spores, micro-algae and more. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24