Learn More
Bertin Corp Micro-organism Lysing VK01 - 2mL tubes 50 Prep
SDP
Catalog No. 501542934
Encompass_Preferred
Lysing kit for Precellys - VK01 Microorganism kit - 2mL tubes 50 prep. The VK01 matrix is composed of 0.1mm glass beads and is designed for micro-organisms lysing like gram- and gram+ bacteria but also spores, micro-algae etc. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24
Catalog No. 50-154-2934 Supplier Bertin Corp Supplier No. P000914LYSK0A.0
Lysing kit for Precellys - VK01 Microorganism kit - 2mL tubes 50 prep. The VK01 matrix is composed of 0.1mm glass beads and is designed for micro-organisms lysing like gram- and gram+ bacteria but also spores, micro-algae etc. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24