Learn More
Bertin Corp Hard Tissue Grinding MK28-R - 2mL Reinforced tubes 50 Prep
SDP
Catalog No. 501542937
Encompass_Preferred
Reinforced Lysing kit for Precellys - MK28-R Hard tissue kit - 2mL tubes 50 prep. The MK28 matrix is composed of 2.8mm Metal beads recommended for hard tissues such as muscle, skin, spinal cord, bone, teeth, hair but also for plant roots, rods or seeds like corn, rice, wheat etc. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24.
Catalog No. 50-154-2937 Supplier Bertin Corp Supplier No. P000917LYSK0A.0
Reinforced Lysing kit for Precellys - MK28-R Hard tissue kit - 2mL tubes 50 prep. The MK28 matrix is composed of 2.8mm Metal beads recommended for hard tissues such as muscle, skin, spinal cord, bone, teeth, hair but also for plant roots, rods or seeds like corn, rice, wheat etc. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24.