Learn More
Bertin Corp Tissue Homogenizing CKMix - 2mL tubes 50 Prep
SDP
Catalog No. 501542938
Encompass_Preferred
Lysing kit for Precellys - CKMix Tissue kit - 2mL tubes 50 prep. The CKMix is a ceramic matrix composed of 2.8mm and 1.4mm beads and is indicated in the homogenization of soft to hard tissues such as liver, kidney, ear, lung, heart, spleen, tail, artery, tumor, intestine, adipose tissue but also on plants parts such as leaves or small roots. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24
Catalog No. 50-154-2938 Supplier Bertin Corp Supplier No. P000918LYSK0A.0
Lysing kit for Precellys - CKMix Tissue kit - 2mL tubes 50 prep. The CKMix is a ceramic matrix composed of 2.8mm and 1.4mm beads and is indicated in the homogenization of soft to hard tissues such as liver, kidney, ear, lung, heart, spleen, tail, artery, tumor, intestine, adipose tissue but also on plants parts such as leaves or small roots. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. Compatible with MP Fastprep, Omni Bead Ruptor BR24