Learn More
Bertin Corp Micro-organism Lysing VKMix - 7mL tubes 50 Prep
SDP
Catalog No. 501542955
Encompass_Preferred
Lysing kit for Precellys - VKMix Micro-Organism kit - 7mL tubes 50 prep. The VKMix matrix is composed of 0.1mm and 0.5mm glass beads and is designed for micro-organisms lysing like gram- and gram+ bacteria but also spores, micro-algae etc. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 200mg to 2g of tissue
Catalog No. 50-154-2955 Supplier Bertin Corp Supplier No. P000937LYSK0A.0
Lysing kit for Precellys - VKMix Micro-Organism kit - 7mL tubes 50 prep. The VKMix matrix is composed of 0.1mm and 0.5mm glass beads and is designed for micro-organisms lysing like gram- and gram+ bacteria but also spores, micro-algae etc. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 200mg to 2g of tissue