Learn More
Bertin Corp Soft Tissue Homogenizing CK14 - 7mL tubes 50 Prep
SDP
Catalog No. 501542958
Encompass_Preferred
Lysing kit for Precellys - CK14 Soft Tissue kit - 7mL tubes 50 prep. The CK14 is a ceramic (zirconium oxide) matrix composed of 1.4mm beads and is indicated in the homogenization of soft tissues such as liver, kidney, brain, but also on plants parts such as leaves. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 200mg to 2g of tissue. The kits requires the use of the dedicated tube holder for Precellys Evolution (SP02520-810-NC010.0)
Catalog No. 50-154-2958 Supplier Bertin Corp Supplier No. P000940LYSK0A.0
Lysing kit for Precellys - CK14 Soft Tissue kit - 7mL tubes 50 prep. The CK14 is a ceramic (zirconium oxide) matrix composed of 1.4mm beads and is indicated in the homogenization of soft tissues such as liver, kidney, brain, but also on plants parts such as leaves. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 200mg to 2g of tissue. The kits requires the use of the dedicated tube holder for Precellys Evolution (SP02520-810-NC010.0)