Learn More
Bertin Corp Sample Grinding GK60 - 2mL Reinforced tubes 50 Prep
SDP
Catalog No. 501542942
Encompass_Preferred
Includes 50 preps of garnet flakes 0.7mm and one ceramic (Zirconium Oxide, Ceria stabilized) in 2mL reinforced tubes. GK60 is designed for wet or dry tissue grinding like liver, plants, or dry samples like rice and more. Sample size range: from 20mg to 200mg tissue or 200L to 1.6mL culture.
Catalog No. 50-154-2942 Supplier Bertin Corp Supplier No. P000924LYSK0A.0
Includes 50 preps of garnet flakes 0.7mm and one ceramic (Zirconium Oxide, Ceria stabilized) in 2mL reinforced tubes. GK60 is designed for wet or dry tissue grinding like liver, plants, or dry samples like rice and more. Sample size range: from 20mg to 200mg tissue or 200L to 1.6mL culture.