Learn More
Bertin Corp Metal tube for Precellys Evolution
SDP
Catalog No. 501542968
Encompass_Preferred
Metal tube is the ideal tool for homogenization of very hard tough samples like bones, teeth, coral etc. A grinding matrix must be filled in the tube before use using beads in bulk available in the Precellys catalog
Catalog No. 50-154-2968 Supplier Bertin Corp Supplier No. P000952LYSK0A.0
Metal tube is the ideal tool for homogenization of very hard tough samples like bones, teeth, coral etc. A grinding matrix must be filled in the tube before use using beads in bulk available in the Precellys catalog