Learn More
Cayman Chemical Precellys Stainless Steel Be
SDP
Catalog No. 502887043
Encompass_Preferred
Includes 500 preps of 2.8 mm metal beads. These beads are designed for use with Precellys Tissue Homogenizers. Beads are designed for hard tissue grinding like muscle, skin, spincor, bone, teeth, hair, seeds, corn, rice, and more. [Bertin Catalog P000925-LYSK0-A.0; Previously KT03961-1-101.BK]
Catalog No. 50-288-7043 Supplier Cayman Chemical Supplier No. 104001 BOX
Includes 500 preps of 2.8 mm metal beads. These beads are designed for use with Precellys Tissue Homogenizers. Beads are designed for hard tissue grinding like muscle, skin, spincor, bone, teeth, hair, seeds, corn, rice, and more. [Bertin Catalog P000925-LYSK0-A.0; Previously KT03961-1-101.BK]