Learn More
Cayman Chemical Precellys Zirconium Oxide Be
SDP
Catalog No. 502886956
Encompass_Preferred
Includes 500 preps of 2.8 mm ceramic beads. These beads are designed for use with Precellys Tissue Homogenizers. Beads are designed for hard tissue homogenization like EAr, lung, heart, spleen, tail, cornea, artery, arabidopsis, leaves, and more. [Bertin Catalog P000926-LYSK0-A.0; Previously KT03961-1-102.BK]
Catalog No. 50-288-6956 Supplier Cayman Chemical Supplier No. 104011 BOX
Includes 500 preps of 2.8 mm ceramic beads. These beads are designed for use with Precellys Tissue Homogenizers. Beads are designed for hard tissue homogenization like EAr, lung, heart, spleen, tail, cornea, artery, arabidopsis, leaves, and more. [Bertin Catalog P000926-LYSK0-A.0; Previously KT03961-1-102.BK]