Learn More
Cayman Chemical Precelys 15 mL Dry Hard TIs 1
SDP
Catalog No. 502965983
Encompass_Preferred
Precellys Dry Hard Tissue Grinding Kit includes 25 preps of 6.8 mm ceramic beads (CK68) in 15 mL tubes. Custom design reinforced tubes for bead beating application: O'ring and reinforced materials for dry grinding, valuable or very hard samples. CK68 is designed for dry samples grinding like rice grains, pepper grains, and more. Sample size range from 400 mg to 4 g tissue or 400 µL to 9 mL culture. [Bertin Catalog P000950-LYSK0-A.0; Previously KT03961-1-514.15]
Catalog No. 50-296-5983 Supplier Cayman Chemical Supplier No. 169141 BOX
Precellys Dry Hard Tissue Grinding Kit includes 25 preps of 6.8 mm ceramic beads (CK68) in 15 mL tubes. Custom design reinforced tubes for bead beating application: O'ring and reinforced materials for dry grinding, valuable or very hard samples. CK68 is designed for dry samples grinding like rice grains, pepper grains, and more. Sample size range from 400 mg to 4 g tissue or 400 µL to 9 mL culture. [Bertin Catalog P000950-LYSK0-A.0; Previously KT03961-1-514.15]