Learn More
Cayman Chemical Precelys 15 mL Hard TIsue 1BOX
SDP
Catalog No. 502966425
Encompass_Preferred
Precellys Hard Tissue Homogenizing Kit includes 25 preps of 2.8 mm ceramic beads (CK28) in 15 ml tubes. CK28 beads are designed for hard tissue homogenization like EAr, lung, heart, spleen, tail, artery, arabidopsis, leaves, and more. Sample size range from 400 mg to 4 g tissue or 400 µl to 9 ml culture. [Bertin Catalog P000947-LYSK0-A.0; Previously KT03961-1-502.15]
Catalog No. 50-296-6425 Supplier Cayman Chemical Supplier No. 169121 BOX
Precellys Hard Tissue Homogenizing Kit includes 25 preps of 2.8 mm ceramic beads (CK28) in 15 ml tubes. CK28 beads are designed for hard tissue homogenization like EAr, lung, heart, spleen, tail, artery, arabidopsis, leaves, and more. Sample size range from 400 mg to 4 g tissue or 400 µl to 9 ml culture. [Bertin Catalog P000947-LYSK0-A.0; Previously KT03961-1-502.15]