Learn More
Cayman Chemical Precelys 2ml Dry Hard TIs 10
SDP
Catalog No. 502969615
Encompass_Preferred
Kit includes 50 preps of ceramic (zirconium oxide) beads of 6.8 mm in 2 mL reinforced tubes. Custom design reinforced tubes for bead beating application: O-ring and reinforced materials for dry grinding, valuable or very hard samples. CK68 Reinforced is designed for dry samples grinding like rice grains, pepper grains and more. Sample size range from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000923-LYSK0-A.0; Previously KT03961-1-014.2]
Catalog No. 50-296-9615 Supplier Cayman Chemical Supplier No. 1523510 BOX
Kit includes 50 preps of ceramic (zirconium oxide) beads of 6.8 mm in 2 mL reinforced tubes. Custom design reinforced tubes for bead beating application: O-ring and reinforced materials for dry grinding, valuable or very hard samples. CK68 Reinforced is designed for dry samples grinding like rice grains, pepper grains and more. Sample size range from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000923-LYSK0-A.0; Previously KT03961-1-014.2]