Learn More
Cayman Chemical Precelys 2ml Empty ReInfor 1
SDP
Catalog No. 502895963
Encompass_Preferred
Precellys Reinforced Tubes (empty) 2 ml are designed for the homogenization, grinding, and lysis on Precellys 24, Precellys Minilys Personal Tissue Homogenizer (Item No. 10435), or Precellys Evolution Touch (Item No. 38707). Includes 200 x 2 ml reinforced tubes and caps. [Bertin Catalog P000943-LYSK0-A.0; Previously KT03961-1-403.2]
Catalog No. 50-289-5963 Supplier Cayman Chemical Supplier No. 90005381 BOX
Precellys Reinforced Tubes (empty) 2 ml are designed for the homogenization, grinding, and lysis on Precellys 24, Precellys Minilys Personal Tissue Homogenizer (Item No. 10435), or Precellys Evolution Touch (Item No. 38707). Includes 200 x 2 ml reinforced tubes and caps. [Bertin Catalog P000943-LYSK0-A.0; Previously KT03961-1-403.2]