Learn More
Cayman Chemical Precelys 2ml GrIndI 1box
SDP
Catalog No. 502965150
Encompass_Preferred
The kit includes 50 preps of garnet flakes 0.7 mm and one ceramic (Zirconium Oxide, Ceria stabilized) in 2 mL reinforced tubes. GK60 is designed for wet or dry tissue grinding like liver, plants, dry samples like rice, and more. Sample size range: from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000924-LYSK0-A.0; Previously KT03961-1-015.2]
Catalog No. 50-296-5150 Supplier Cayman Chemical Supplier No. 179521 BOX
The kit includes 50 preps of garnet flakes 0.7 mm and one ceramic (Zirconium Oxide, Ceria stabilized) in 2 mL reinforced tubes. GK60 is designed for wet or dry tissue grinding like liver, plants, dry samples like rice, and more. Sample size range: from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000924-LYSK0-A.0; Previously KT03961-1-015.2]