Learn More
Cayman Chemical Precelys 7 mL MIcroorgnIs 1BOX
SDP
Catalog No. 502955331
Encompass_Preferred
Kit includes 50 preps of glass mix beads of 0.1 mm and 0.5 mm in 7 ml standard tubes with O-ring. VKMix 7 ml is designed for micro-organism lysing like yeast fungi, gram+, gram- bacteria, spores, micro-algae and more. Sample size range from 200 mg to 2 g sample or 200 µl to 6 ml culture. [Bertin Catalog P000937-LYSK0-A.0; Previously KT03961-1-304.7]
Catalog No. 50-295-5331 Supplier Cayman Chemical Supplier No. 137631 BOX
Kit includes 50 preps of glass mix beads of 0.1 mm and 0.5 mm in 7 ml standard tubes with O-ring. VKMix 7 ml is designed for micro-organism lysing like yeast fungi, gram+, gram- bacteria, spores, micro-algae and more. Sample size range from 200 mg to 2 g sample or 200 µl to 6 ml culture. [Bertin Catalog P000937-LYSK0-A.0; Previously KT03961-1-304.7]