Learn More
Cayman Chemical Precelys 7 mL Tube Rack 1ea
SDP
Catalog No. 502954542
Encompass_Preferred
Easily store and transport the large 7 mL tubes designed for use with the Precellys line of homogenizers. The 7 mL tube rack can accommodate up to 12 x 7 mL tubes. [Bertin Catalog S000748-PR480-A.0; Previously SP03961-810-NC001.0]
Catalog No. 50-295-4542 Supplier Cayman Chemical Supplier No. 139351 EA
Easily store and transport the large 7 mL tubes designed for use with the Precellys line of homogenizers. The 7 mL tube rack can accommodate up to 12 x 7 mL tubes. [Bertin Catalog S000748-PR480-A.0; Previously SP03961-810-NC001.0]