Learn More
Cayman Chemical Precelys EvolutIon 15 mL 1ea
SDP
Catalog No. 502968121
Encompass_Preferred
Includes 50 units of 15 mL tubes and caps, assembled. Empty 15 mL tubes are designed for 15 mL homogenization, grinding, and lysing on Precellys Evolution Touch (Item No. 38707). . Beads have to be added. [Bertin Catalog P000946-LYSK0-A.0; Previously KT03961-1-406.15]
Catalog No. 50-296-8121 Supplier Cayman Chemical Supplier No. 168691 EA
Includes 50 units of 15 mL tubes and caps, assembled. Empty 15 mL tubes are designed for 15 mL homogenization, grinding, and lysing on Precellys Evolution Touch (Item No. 38707). . Beads have to be added. [Bertin Catalog P000946-LYSK0-A.0; Previously KT03961-1-406.15]