Learn More
Cayman Chemical Precelys EvolutIon 15 mL 1ea
SDP
Catalog No. 502988176
Encompass_Preferred
The 15 mL Blue Blocking Plate is an accessory for the Precellys Evolution Touch (Item No. 38707) and must be used in conjunction with the 15 mL Tube Holder, which is included in the Precellys Evolution - 15 mL Holder Pack (Item No. 16908). This accessory blocks 6 x 15 mL Precellys tubes. [Bertin Catalog S000804-PEVO0-A.0]
Catalog No. 50-298-8176 Supplier Cayman Chemical Supplier No. 301261 EA
The 15 mL Blue Blocking Plate is an accessory for the Precellys Evolution Touch (Item No. 38707) and must be used in conjunction with the 15 mL Tube Holder, which is included in the Precellys Evolution - 15 mL Holder Pack (Item No. 16908). This accessory blocks 6 x 15 mL Precellys tubes. [Bertin Catalog S000804-PEVO0-A.0]