Learn More
Cayman Chemical Precelys EvolutIon 7 mL1BOX
SDP
Catalog No. 502954290
Encompass_Preferred
Includes 50 units of 7 mL tubes and caps, assembled. Empty 7 mL tubes are designed for 7 mL homogenization, grinding, and lysing on Precellys Evolution Touch (Item No. 38707). Beads have to be added. Also compatible with the Precellys Minilys Personal Tissue Homogenizer (Item No. 10435). [Bertin Catalog P000944-LYSK0-A.0; Previously KT03961-1-404.7]
Catalog No. 50-295-4290 Supplier Cayman Chemical Supplier No. 139171 BOX
Includes 50 units of 7 mL tubes and caps, assembled. Empty 7 mL tubes are designed for 7 mL homogenization, grinding, and lysing on Precellys Evolution Touch (Item No. 38707). Beads have to be added. Also compatible with the Precellys Minilys Personal Tissue Homogenizer (Item No. 10435). [Bertin Catalog P000944-LYSK0-A.0; Previously KT03961-1-404.7]