Learn More
Cayman Chemical Precelys EvolutIon 96Wel 1ea
SDP
Catalog No. 502984721
Encompass_Preferred
Package Includes: 1 plate with 12 strips of 8 wells, EAch well prefilled with a mix of ceramic beads. Includes 12 x caps for strips.Homogenize or lyse 96 tissue, plant, or cell samples in prefilled 300 µl wells in a mix of ceramic beads. Maximum sample volume is 120 µl, and maximum sample weight is 12 mg. Plates require both the 96-Well Plate Blocking Plate (Item No. 19983), and 96-Well Plate Holder (Item No. 19982) to be used with the Precellys Evolution. Maximum speed and cycles is 8,500 rpm, at 3 x 30 seconds. [Bertin Catalog P000959-LYSK0-A.0; Previously KT03961-1-709.WP]
Catalog No. 50-298-4721 Supplier Cayman Chemical Supplier No. 199811 EA
Package Includes: 1 plate with 12 strips of 8 wells, EAch well prefilled with a mix of ceramic beads. Includes 12 x caps for strips.Homogenize or lyse 96 tissue, plant, or cell samples in prefilled 300 µl wells in a mix of ceramic beads. Maximum sample volume is 120 µl, and maximum sample weight is 12 mg. Plates require both the 96-Well Plate Blocking Plate (Item No. 19983), and 96-Well Plate Holder (Item No. 19982) to be used with the Precellys Evolution. Maximum speed and cycles is 8,500 rpm, at 3 x 30 seconds. [Bertin Catalog P000959-LYSK0-A.0; Previously KT03961-1-709.WP]