Learn More
Cayman Chemical Precelys EvolutIon 96Wel 1ea
SDP
Supplier:  Cayman Chemical 199841 EA
Encompass_Preferred
The Precellys Evolution - 96-Well Plate Tubes Holder is an accessory for the Precellys Evolution Touch (Item No. 38707) and must be used in conjunction with the Precellys Evolution - 96-Well Plate Blocking Plate (Item No. 19983). This accessory holds 1 x Precellys - 96-Well (ceramic) Bead Mix Plate Kit (Item No. 19981). [Bertin Catalog S000814-PEVO0-A.0; Previously SP02520-820-NC013.0]
Catalog No. 50-298-2876