Learn More
Cayman Chemical Precelys EvolutIon 96Wel 1ea
SDP
Supplier:  Cayman Chemical 199821 EA
Encompass_Preferred
Package Includes: 1 x Precellys Evoltuion - 96-Well Plate Blocking Plate (Item No. 19983), 1 x Precellys Evolution - 96-Well Plate Tubes Holder (Item No. 19984) The Precellys - 96-Well Plate Holder Pack contains everything a Precellys Evolution user needs to run the Precellys - 96-Well (ceramic) Bead Mix Plate Kit (Item No. 19981). This product is NOT compatible with the Precellys 24 OR Precellys Minilys. [Bertin Catalog S000811-PEVO0-A.0; Previously SP02520-810-NC012.0]
Catalog No. 50-298-3770