Learn More
Cayman Chemical Precelys EvolutIon ContaI 1ea
SDP
Catalog No. 502967308
Encompass_Preferred
The Containment Seal replacement part is only for use with the Precellys Evolution Touch (Item No. 38707). If the homogenizer is still under warranty contact our technical service department for support. [Bertin Catalog S000805-PEVO0-A.0; Previously SP02520-810-NC004.0]
Catalog No. 50-296-7308 Supplier Cayman Chemical Supplier No. 169111 EA
The Containment Seal replacement part is only for use with the Precellys Evolution Touch (Item No. 38707). If the homogenizer is still under warranty contact our technical service department for support. [Bertin Catalog S000805-PEVO0-A.0; Previously SP02520-810-NC004.0]