Learn More
Cayman Chemical Precelys EvolutIon StarTR 1ea
SDP
Catalog No. 502967440
Encompass
A cyclic oligosaccharide containing seven D-( )-glucopyranose units that is widely used to improve the aqueous solubility of various compounds especially those containing a phenyl group
Catalog No. 50-296-7440 Supplier Cayman Chemical Supplier No. 169011 EA
A cyclic oligosaccharide containing seven D-( )-glucopyranose units that is widely used to improve the aqueous solubility of various compounds especially those containing a phenyl group