Learn More
Cayman Chemical Precelys MInIlys MIcrotub 1ea
SDP
Catalog No. 502954076
Encompass_Preferred
The Minilys Microtube Adapter Set has been specially designed to allow users to process up to 3 x 1.5 mL micro-tubes using the Minilys. [Bertin Catalog S000688-MLYS0-A.0; Previously SP06404-810-NC007.0]
Catalog No. 50-295-4076 Supplier Cayman Chemical Supplier No. 139371 EA
The Minilys Microtube Adapter Set has been specially designed to allow users to process up to 3 x 1.5 mL micro-tubes using the Minilys. [Bertin Catalog S000688-MLYS0-A.0; Previously SP06404-810-NC007.0]